Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLI4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554703
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554703 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554703 Supplier Invitrogen™ Supplier No. PA554703
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IQESPSVPSP VSLSSPGTPG TQHHEPQLHL HGHQHGSPGS SPKVLSQPSD LDLQDVEEVE IGR Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 61%.

GLI4 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLI4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gli4
Gene Accession No. P10075
Gene Alias 9130023H24Rik; GLI family zinc finger 4; GLI4; GLI-Kruppel family member GLI4; GLI-Kruppel family member GLI4 (oncogene HKR4); glioma-associated oncogene family zinc finger 4; HKR4; Krueppel-related zinc finger protein 4; Protein HKR4; RIKEN cDNA 9130023H24 gene; zinc finger protein GLI4; ZNF928
Gene Symbols Gli4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GLI4. Recombinant protein control fragment (Product #RP-93663).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2738
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.