Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLI4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982842
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2842 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2842 Supplier Bioss Supplier No. BS13373R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GLI4 is a protein coding gene.

Specifications

Antigen GLI4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Gli4
Gene Accession No. P10075
Gene Alias 9130023H24Rik; GLI family zinc finger 4; GLI4; GLI-Kruppel family member GLI4; GLI-Kruppel family member GLI4 (oncogene HKR4); glioma-associated oncogene family zinc finger 4; HKR4; Krueppel-related zinc finger protein 4; Protein HKR4; RIKEN cDNA 9130023H24 gene; Zinc finger protein GLI4; ZNF928
Gene Symbols Gli4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GLI4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2738
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.