Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gliadin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982843
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2843 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2843 Supplier Bioss Supplier No. BS13374R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Celiac disease is associated with a CD4+ T-cell response to epitopes of gliadin presented by HLA-DQ2 or -DQ8 class II MHC molecules. These epitopes are present in a 33-mer peptide of wheat alpha-gliadin, residues 56-88, which is resistant to digestion and forms a substrate for tissue transglutaminase (TG2), generating the glutamic acid residues essential for binding to HLA-DQ2. Complete homology exists between residues 63-73, 70-80 and 77-87 of wheat alpha gliadin.

Specifications

Antigen Gliadin
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Alias AGP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from wheat Gliadin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.