Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Glial Fibrillary Acidic Protein Cocktail, Mouse, Unlabeled, Clone: SMI-22,

Catalog No. NE1015100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NE1015100UL 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NE1015100UL Supplier MilliporeSigma™ Supplier No. NE1015100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recognizes ∽50kDa glial fibrillary acidic protein (GFAP) in human and bovine cytoskeletal preparations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glial Fibrillary Acidic Protein Cocktail
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone SMI-22
Conjugate Unconjugated
Formulation Undiluted ascites, ≤ 0.1% Sodium Azide.
Gene Alias Anti-GFAP Cocktail
Host Species Mouse
Immunogen purified bovine GFAP protein
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bovine
Content And Storage -20°C
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.