Learn More
Sigma Aldrich Glider Grides Standard Copper/palladium Square Mesh 200 Mesh 100/vial
Catalog No. 503125556 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 Pk.
Catalog No. Quantity
50-312-5556 1 Pk.
1 options
Catalog No. 50-312-5556 Supplier Sigma Aldrich Supplier No. TEMG200CP
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Gilder Grids Standard Square Mesh: Copper/Palladium Grid, 200 mesh, 3.05mm diameter, 0.7 mil (18mum) thickness. Pitch 125mum, Hole 90mum, Bar 35mum
Specifications
Quantity 1 Pk.
Synonym SQUARE MESH CU/PD 100 VIAL