Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
        Glipizide, USP, 98-102%, Spectrum™ Chemical
 
    
    
    
        
            Shop All Spectrum Chemical Manufacturing Corporation Products
        
    
    
    
    
    
    
    
    
        
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                1 g
                            
                        
                            
                                5 g
                            
                        
                            
                                25 g
                            
                        
                            
                                100 g
                            
                        
                            
                                1 kg
                            
                        
                    
                            
                                Packaging:
                            
                        
                        
                            
                                Amber Glass Bottle
                            
                        
                    Description
Spectrum™ Chemical Glipizide, USP  is an oral diabetes medicine that helps control blood sugar levels by helping your pancreas produce insulin. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
                        
                        
                        
                        
                        
                        
                        
                        
                        
                    
                
                
                    
                
                
    
                
    
            Specifications
Specifications
| CAS Max % | 100% | 
| Ignition Residue | 0.4% | 
| Loss on Drying | 1% | 
| Packaging | Amber Glass Bottle | 
| Quantity | 1 kg | 
| Percent Purity | 98 to 102% | 
| Grade | USP | 
| Physical Form | Crystalline Powder or Lumps | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                                