Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLIPR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552774
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552774 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552774 Supplier Invitrogen™ Supplier No. PA552774
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NEDFIKDCVR IHNKFRSEVK PTASDMLYMT WDPALAQIAK AWASNCQFSH NTRLKPPHKL HPNFTSLGEN IWTGSVPIFS VSSAITNWYD EIQDYDF Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 61%.

The glioma pathogensis-related protein 1 (GLIPR1) is similar to both the pathogenesis-related protein (PR) superfamily and the cysteine-rich secretory protein (CRISP) family. GLIPR1 is a tumor suppressor whose expression is regulated by p53; its increased expression is associated with myelomocytic differentiation in macrophages, whereas decreased expression of this gene through gene methylation is associated with prostate cancer. The GLIPR1 gene is part of a p53 target gene cluster that includes GLIPR1L1 and GLIPR1L2, two highly homologous proteins whose expression patterns vary.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLIPR1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GLIPR1
Gene Accession No. P48060
Gene Alias 2410114O14Rik; CRISP7; GLI pathogenesis related 1; GLI pathogenesis-related 1; GLI pathogenesis-related 1 (glioma); glioma pathogenesis-related protein 1; GLIPR; gliPR 1; Glipr1; mRTVP-1; protein RTVP-1; related to testes-specific, vespid, and pathogenesis proteins; related to testis-specific, vespid, and pathogenesis proteins 1; RTVP1; RTVP-1; testes-specific vespid and pathogenesis protein 1
Gene Symbols GLIPR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GLIPR1. Recombinant protein control fragment (Product #RP-90273).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11010
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.