Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GLIPR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595776
Description
Immunogen sequence: ANILPDIENE DFIKDCVRIH NKFRSEVKPT ASDMLYMTWD PALAQIAKAW ASNCQFSHNT RLKPPHKLHP NFTSLGENIW TGSVPIFSVS SAITNWYDEI QDYDFKTRIC KKVCGHYTQ; Positive Samples: Rat lung; Cellular Location: Membrane, Single-pass membrane protein.
The glioma pathogensis-related protein 1 (GLIPR1) is similar to both the pathogenesis-related protein (PR) superfamily and the cysteine-rich secretory protein (CRISP) family. GLIPR1 is a tumor suppressor whose expression is regulated by p53; its increased expression is associated with myelomocytic differentiation in macrophages, whereas decreased expression of this gene through gene methylation is associated with prostate cancer. The GLIPR1 gene is part of a p53 target gene cluster that includes GLIPR1L1 and GLIPR1L2, two highly homologous proteins whose expression patterns vary.
Specifications
GLIPR1 | |
Polyclonal | |
Unconjugated | |
GLIPR1 | |
GLIPR1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 22-140 of human GLIPR1 (NP_0068422). | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
1.35 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
2410114O14Rik; CRISP7; GLI pathogenesis related 1; GLI pathogenesis-related 1; GLI pathogenesis-related 1 (glioma); glioma pathogenesis-related protein 1; GLIPR; gliPR 1; Glipr1; mRTVP-1; protein RTVP-1; related to testes-specific, vespid, and pathogenesis proteins; related to testis-specific, vespid, and pathogenesis proteins 1; RTVP1; RTVP-1; testes-specific vespid and pathogenesis protein 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
299783 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction