Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLIPR1L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528524
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528524 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528524 Supplier Invitrogen™ Supplier No. PA528524
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, PC-3, U87-MG. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Plays a role in the binding between sperm and oocytes. Component of epididymosomes, one type of membranous microvesicules which mediate the transfer of lipids and proteins to spermatozoa plasma membrane during epididymal maturation. Also component of the CD9-positive microvesicules found in the cauda region. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLIPR1L1
Applications Western Blot
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene GLIPR1L1
Gene Accession No. Q6UWM5
Gene Alias ALKN2972; GLI pathogenesis related 1 like 1; GLI pathogenesis-related 1 like 1; GLIPR1L1; GLIPR1-like protein 1; PRO7434; UNQ2972/PRO7434
Gene Symbols GLIPR1L1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GLIPR1L1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256710
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.