Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLIPR1L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559865
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559865 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559865 Supplier Invitrogen™ Supplier No. PA559865
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SKIPSITDPH FIDNCIEAHN EWRGKVNPPA ADMKYMIWDK GLAKMAKAWA NQCKFEHNDC LDKSYKCYAA FEYVGENIWL GGIKS Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 47%.

Plays a role in the binding between sperm and oocytes. Component of epididymosomes, one type of membranous microvesicules which mediate the transfer of lipids and proteins to spermatozoa plasma membrane during epididymal maturation. Also component of the CD9-positive microvesicules found in the cauda region. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLIPR1L1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GLIPR1L1
Gene Accession No. Q6UWM5
Gene Alias ALKN2972; GLI pathogenesis related 1 like 1; GLI pathogenesis-related 1 like 1; GLIPR1L1; GLIPR1-like protein 1; PRO7434; UNQ2972/PRO7434
Gene Symbols GLIPR1L1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GLIPR1L1. Recombinant protein control fragment (Product #RP-97830).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256710
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.