Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLIPR1L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558956
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558956 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558956 Supplier Invitrogen™ Supplier No. PA558956
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QSQFPNILLE QQMIFTPEES EAGNEEEE Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 43%.

This gene encodes a member of the cysteine-rich secretory protein, antigen 5, and pathogenesis-related 1 superfamily. Members of this family have roles in a variety of processes, including cancer and immune defense. This gene is located in a cluster with two related genes on chromosome 12. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLIPR1L2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GLIPR1L2
Gene Accession No. Q4G1C9
Gene Alias GLI pathogenesis related 1 like 2; GLI pathogenesis-related 1 like 2; GLIPR1 like 2; GLIPR1L2; GLIPR1-like protein 2; RGD1310205
Gene Symbols GLIPR1L2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GLIPR1L2. Recombinant protein control fragment (Product #RP-97861).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144321
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.