Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLIPR1L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572789
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572789 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572789 Supplier Invitrogen™ Supplier No. PA572789
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GLIPR1L2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a member of the cysteine-rich secretory protein, antigen 5, and pathogenesis-related 1 superfamily. Members of this family have roles in a variety of processes, including cancer and immune defense. This gene is located in a cluster with two related genes on chromosome 12. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLIPR1L2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene GLIPR1L2
Gene Accession No. Q4G1C9
Gene Alias GLI pathogenesis related 1 like 2; GLI pathogenesis-related 1 like 2; GLIPR1 like 2; GLIPR1L2; GLIPR1-like protein 2; RGD1310205
Gene Symbols GLIPR1L2
Host Species Rabbit
Immunogen GLIPR1L2 antibody for an 15 amino acid peptide near the center of human GLIPR1L2. The immunogen is within amino acids 120 - 170 of GLIPR1L2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144321
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.