Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLIPR1L2 Polyclonal Antibody
GLIPR1L2 Polyclonal Antibody
Supplier: Thermo Scientific PA546655
Description
GLIPR1L2 Polyclonal Antibody for Western Blot

Specifications
GLIPR1L2 | |
Polyclonal | |
Unconjugated | |
GLIPR1L2 | |
GLI pathogenesis related 1 like 2; GLI pathogenesis-related 1 like 2; GLIPR1 like 2; GLIPR1L2; GLIPR1-like protein 2; RGD1310205 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
144321 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q4G1C9 | |
GLIPR1L2 | |
Synthetic peptide directed towards the C-terminal of human GLIPR1L2 (aa 288-337). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction