Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLIPR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556164
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556164 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556164 Supplier Invitrogen™ Supplier No. PA556164
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GKSASKQFHN EVLKAHNEYR QKHGVPPLKL CKNLNREAQQ YSEALASTRI LKHSPESSRG QCGENLAWAS YD Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 90%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen GLIPR2
Applications Western Blot
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GLIPR2
Gene Accession No. Q9H4G4
Gene Alias 17kD fetal brain protein; 5730414A08Rik; C77180; C9orf19; GAPR1; GAPR-1; GLI pathogenesis related 2; GLI pathogenesis-related 2; glioma pathogenesis-related protein 2; GliPR 2; Glipr2; Golgi-associated plant pathogenesis-related protein 1; golgi-associated PR-1 protein
Gene Symbols GLIPR2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GLIPR2. Recombinant protein control fragment (Product #RP-92935).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152007
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.