Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLIS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568892
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568892 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568892 Supplier Invitrogen™ Supplier No. PA568892
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 79%; Dog: 91%; Mouse: 100%; Pig: 79%; Rabbit: 86%; Rat: 93%.

Glis1 (Gli similar 1) is also known as Gli related Kruppel-like zinc finger 1 and is encoded by the Glis1 gene located on chromosome 1 in humans. It belongs to the Kruppel-like zinc finger family of proteins that constitute mainly of transcription factors and is hence studied for its role in target gene activation and repression, specifically during embryonic development. Glis1 contains five tandem zinc finger motifs, which is the region at which it shares close homology to Glis2 and Glis 3. More recent studies on Glis1 show its interaction with Oct 3/4, Sox2 and KLF4 in efficient generation of iPSC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLIS1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GLIS1
Gene Accession No. Q8K1M4
Gene Alias gli homologous protein 1; Gli5; Gli6; GliH1; GLI-Kruppel family member GLI6; GLIS family zinc finger 1; GLIS1; Glis1 Kruppel-like zinc finger protein; GLI-similar 1; translation initiation factor IF-2; zinc finger protein GLIS1; zinc finger transcription factor Gli5
Gene Symbols GLIS1
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse GLIS1 (aa 11-60).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230587
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.