Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLIS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140108
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140108 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140108 Supplier Invitrogen™ Supplier No. PA5140108
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: VEGRFSAAPL VDLSLSPPSG LDSPNGSSSL SPERQGNGDL PPVPSASDFQ PLRYLDGVPS S Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 92%.

Members of the Kruppel-like zinc finger protein family, such as GLIS2, function as activators and/or repressors of gene transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLIS2
Applications Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GLIS2
Gene Accession No. Q9BZE0
Gene Alias Gli5; GLI-Kruppel family member GLI5; GLIS family zinc finger 2; GLIS2; GLI-similar 2; Klf16; Kruppel-like factor 16 (neuronal); Kruppel-like zinc finger protein GLIS2; nephrocystin-7; neuronal Krueppel-like protein; NKL; NPHP7; Zinc finger protein GLI5; zinc finger protein GLIS2
Gene Symbols GLIS2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GLIS2. Recombinant protein control fragment (Product #RP-109026).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84662
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.