Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLIS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541677
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541677 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541677 Supplier Invitrogen™ Supplier No. PA541677
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PPASQVSTAC NQISPSLQRA MNAANLNIPP SDTRSLISRE SLASTTLSLT Sequence homology: Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.

This gene is a member of the GLI-similar zinc finger protein family and encodes a nuclear protein with five C2H2-type zinc finger domains. This protein functions as both a repressor and activator of transcription and is specifically involved in the development of pancreatic beta cells, the thyroid, eye, liver and kidney. Mutations in this gene have been associated with neonatal diabetes and congenital hypothyroidism (NDH). Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only two have been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLIS3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GLIS3
Gene Accession No. Q8NEA6
Gene Alias 4833409N03Rik; E330013K21Rik; GLIS family zinc finger 3; Glis3; GLI-similar 3; NDH; OK/KNS-cl.4; RGD1565505; Zinc finger protein 515; zinc finger protein GLIS3; ZNF515
Gene Symbols GLIS3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human GLIS3 (aa 11-60).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 169792
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.