Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLIS3 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30512820UL
Description
GLIS3 Polyclonal specifically detects GLIS3 in Human, Mouse samples. It is validated for Western Blot.Specifications
GLIS3 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
169792 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
FLJ38999, FLJ90578, GLIS family zinc finger 3, GLI-similar 3, OK/KNS-cl.4, zinc finger protein 515, zinc finger protein GLIS3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 606-775 of human GLIS3 (NP_689842.3). | |
20 μL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction