Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLMN Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297214MFV450
Description
GLMN Polyclonal antibody specifically detects GLMN in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
GLMN | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
FAP, FAP48FAP68, FKBPAP, GLMLFK506-binding protein-associated protein, glomulin, glomulin, FKBP associated protein, GVM, GVMFKBP-associated protein, VMGLOMvenous malformation with glomus cells | |
Produced in rabbits immunized with E. coli-derived Human GLMN fragment. | |
0.1 mL | |
Angiogenesis | |
11146 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction