Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLMN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588301
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588301 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588301 Supplier Invitrogen™ Supplier No. PA588301
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RCLLNTSNHS GVEAFIIQNI KNQIDMSLKR TRNNKWFTGP QLISLLDLVL FLPEGAETDL LQNSDRIMAS LNLLRYLVIK DNENDNQTGL WTELGNIENN FLKPLHIGLN MSKAHYEAEI KNSQEAQKSK DLCSITVSGE EIPNMPPEMQ LKVLHSALFT FDLIESVLAR VEELIEIKTK STSEENIGIK; Positive Samples: A-549, U-87MG, HepG2, Mouse testis.

This gene encodes a phosphorylated protein that is a member of a Skp1-Cullin-F-box-like complex. The protein is essential for normal development of the vasculature and mutations in this gene have been associated with glomuvenous malformations, also called glomangiomas. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLMN
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GLMN
Gene Accession No. Q8BZM1, Q92990
Gene Alias 9330160J16Rik; AW227515; FAP; Fap48; FAP68; FK506-binding protein-associated protein; FKBP associated protein; FKBPAP; FKBP-associated protein; GLML; Glmn; Glomulin; glomulin, FKBP associated protein; glomulin-like protein; GVM; LOW QUALITY PROTEIN: glomulin; VMGLOM
Gene Symbols GLMN
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 405-594 of human GLMN (NP_4445041).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11146, 170823
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.