Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLO1/Glyoxalase I Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501570039
Encompass_Preferred
Change view
Click to view available options
20 μL; Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-157-0039 20 μL; Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-0039 Supplier Bethyl Laboratories, Inc Supplier No. A305290AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

GLO1 is a member of the glyoxalase I family that is responsible for the catalysis and formation of S-lactoyl-glutathione from methylglyoxal condensation and reduced glutathione. It is believed that inactivation of GLO1 in endothelial cells contributes to vascular complications in diabetes. GLO1 has also shown to extend lifespan in C. elegans, and has been shown to be upregulated in maligt cells.

Specifications

Antigen GLO1/Glyoxalase I
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene GLO1
Gene Accession No. Q04760, Q9CPU0
Gene Alias Glo1, Ketone-aldehyde mutase, Aldoketomutase, Methylglyoxalase
Gene Symbols Glo1
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109801, 2739
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.