Learn More
GLO1/Glyoxalase I Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305291AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
GLO1 is a member of the glyoxalase I family that is responsible for the catalysis and formation of S-lactoyl-glutathione from methylglyoxal condensation and reduced glutathione. It is believed that inactivation of GLO1 in endothelial cells contributes to vascular complications in diabetes. GLO1 has also shown to extend lifespan in C. elegans, and has been shown to be upregulated in maligt cells.Specifications
GLO1/Glyoxalase I | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q04760 | |
GLO1 | |
Between 134 and 184 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
GLO1 | |
Glo1, Ketone-aldehyde mutase, Aldoketomutase, Methylglyoxalase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2739 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.