Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581397
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581397 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581397 Supplier Invitrogen™ Supplier No. PA581397
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Mouse Glyoxalase I/GLO1.

The enzyme encoded by this gene is responsible for the catalysis and formation of S-lactoyl-glutathione from methylglyoxal condensation and reduced glutathione. Glyoxalase I is linked to HLA and is localized to 6p21.3-p21.1, between HLA and the centromere.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLO1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene GLO1
Gene Accession No. Q9CPU0
Gene Alias 0610009E22Rik; 1110008E19Rik; 2510049H23Rik; aldoketomutase; AW550643; cb554; epididymis secretory protein Li 74; GLO1; Glo-1; Glo-1r; Glo1-r; Glo-1s; Glo1-s; GLOD1; glx I; GLY1; GLYI; glyoxalase 1; glyoxalase 1 complex; glyoxalase 1 regulatory; glyoxalase 1 structural; glyoxalase domain containing 1; glyoxalase I; glyoxylase 1; HEL-S-74; ketone-aldehyde mutase; lactoyl glutathione lyase; lactoylglutathione lyase; lactoylglutathione lyase GLO1; methylglyoxalase; Qglo; S-D-lactoylglutathione methylglyoxal lyase; wu:fb82g09; YM9571.15C; YML004C; zgc:66035
Gene Symbols GLO1
Host Species Rabbit
Immunogen Recombinant Mouse GLO1 protein (Ala2-Ile184).
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109801
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.