Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595997
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595997 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595997 Supplier Invitrogen™ Supplier No. PA595997
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAEPQPPSGG LTDEAALSCC SDADPSTKDF LLQQTMLRVK DPKKSLDFYT RVLGMTLIQK CDFPIMKFSL YFLAYEDKND IPKEKDEKIA WALSRKATLE LTHNWGTEDD ETQSYHNGNS DPRGFGHIGI AVPDVYSACK RFEELGVKFV KKPDDGKMKG LAFIQDPDGY WIEILNPNKM ATLM; Positive Samples: BT-474, 22Rv1, U-937, PC-3, Mouse kidney.

The enzyme encoded by this gene is responsible for the catalysis and formation of S-lactoyl-glutathione from methylglyoxal condensation and reduced glutathione. Glyoxalase I is linked to HLA and is localized to 6p21.3-p21.1, between HLA and the centromere.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLO1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GLO1
Gene Accession No. Q04760, Q9CPU0
Gene Alias 0610009E22Rik; 1110008E19Rik; 2510049H23Rik; aldoketomutase; AW550643; cb554; epididymis secretory protein Li 74; GLO1; Glo-1; Glo-1r; Glo1-r; Glo-1s; Glo1-s; GLOD1; glx I; GLY1; GLYI; glyoxalase 1; glyoxalase 1 complex; glyoxalase 1 regulatory; glyoxalase 1 structural; glyoxalase domain containing 1; glyoxalase I; glyoxylase 1; HEL-S-74; ketone-aldehyde mutase; lactoyl glutathione lyase; lactoylglutathione lyase; lactoylglutathione lyase GLO1; methylglyoxalase; Qglo; S-D-lactoylglutathione methylglyoxal lyase; wu:fb82g09; YM9571.15C; YML004C; zgc:66035
Gene Symbols GLO1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-184 of human GLO1 (NP_0066992).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109801, 2739
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.