Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Universal Snap Caps

Catalog No. 22171656
Change view
Click to view available options
Color:
Black
Blue
Gray
Green
Lavender
Natural
Red
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-171-656 Blue
22-171-655 Gray
22-171-660 Lavender
22-171-659 Black
22-171-658 Green
22-171-657 Natural
22-171-662 Yellow
22-171-661 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 22-171-656 Supplier Globe Scientific Supplier No. 118115B
Add to Cart
Add to Cart

Provides a leak-resistant closure

  • Precision molded low-density polyethylene made
  • Fits most 12, 13 and 16mm plastic test tubes
  • Avoids cross-contamination, protects lab work areas against infection and spillage
  • Not autoclavable
  • Assorted colors available for sample identification

Specifications

Product Type Snap Cap
Autoclavable Not Autoclavable
Material Polyethylene
Color Blue
Size Universal
For Use With (Equipment) Recapping 12, 13 and 16 mm evacuated glass blood collection test tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.