Learn More
Invitrogen™ Globo H Monoclonal Antibody (VK9), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ 14970095

Description
The VK9 MAb recognizes human Globo H, a hexasaccharide formed from the pentasaccharide precursor, SSEA3 (stage-specific embryonic antigen 3). It is a member of a family of antigenic carbohydrates expressed on the surface of cancer cells as glycolipids and possibly glycoproteins. Globo H is overexpressed on many carcinomas, such as breast, colon, ovarian, gastric, pancreatic, lung, and prostate. Expression is observed in more than half of all ductal, lobular, and tubular breast carcinomas, but not in non-epithelial breast tumors. Globo H is also expressed on cancer stem cells and human embryonic stem cells, although expression decreases when cells are differentiated toward both neural progenitor cells and to a lesser extent in endodermal lineage cells. Expression in normal tissues is limited to cells composing secretory borders. Globo H is considered a target for immunotherapy of many epithelial cancers.
Globo H is a hexasaccharide formed from the pentasaccharide precursor, SSEA3 (stage-specific embryonic antigen 3). It is a member of a family of antigenic carbohydrates expressed on the surface of cancer cells as glycolipids and possibly glycoproteins. Globo H is overexpressed on many carcinomas, such as breast, colon, ovarian, gastric, pancreatic, lung, and prostate. Expression is observed in more than half of all ductal, lobular, and tubular breast carcinomas, but not in non-epithelial breast tumors. Globo H is also expressed on cancer stem cells and human embryonic stem cells, although expression decreases when cells are differentiated toward both neural progenitor cells and to a lesser extent in endodermal lineage cells. Expression in normal tissues is limited to cells composing secretory borders. Globo H is considered a target for immunotherapy of many epithelial cancers.
Specifications
Globo H | |
Monoclonal | |
0.5 mg/mL | |
PBS with proprietary stabilizer and 0.09% sodium azide; pH 7.2 | |
Mouse | |
10 mg | |
Primary | |
4°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
VK9 | |
Unconjugated | |
Globo H antigen; globoH; Globohexaosylceramide | |
Affinity chromatography | |
RUO | |
Human | |
Antibody | |
IgG3 κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.