Learn More
GLOD4 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305664AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Germinal center kinases, such as TNIK, are characterized by an N-terminal kinase domain and a C-terminal GCK domain that serves a regulatory function.Specifications
GLOD4 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9HC38 | |
GLOD4 | |
Between 263 and 313 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
GLOD4 | |
CGI-150, C17orf25, HC71, glyoxalase domain-containing protein 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51031 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.