Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Glove Box Gloves, Sleeves and Hands
SDP

Catalog No. 161013 Shop All Labconco Corporation Products
Change view
Click to view available options
Material:
Butyl
CSM
Neoprene
Size:
10
7.5
8.5
9
9.75
Universal
Thickness (English):
15 mil
17 mil
Color:
Black
White
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Size Thickness (English) Color
16-101-3 Neoprene 7.5 17 mil Black
16-200-23 Butyl 8.5 15 mil Black
16-200-21 Butyl 9.75 15 mil Black
16-200-22 CSM 8.5 15 mil White
16-200-20 CSM 9.75 15 mil White
16-200-2 Neoprene 9.75 15 mil Black
16-101-4 Neoprene 9 17 mil Black
16-200-15 Neoprene 10 17 mil Black
16-200-5 Neoprene Universal 15 mil Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 16-101-3 Supplier Labconco™ Supplier No. 5006000
Add to Cart
Add to Cart
This item is not returnable. View return policy

Glove box gloves, sleeves and hands are available in a variety of sizes, materials and thicknesses to suit your specific application. Choose from neoprene, latex, CSM or butyl.

Glove Box Gloves - One pair/pack, one-piece, seamless, with O-rings

  • Neoprene - Size 9.75
    • 17 mil thickness
  • CSM (Chlorosulfonated polyethylene rubber) - Available in sizes 8.5 or 9.75
    • Also known as Hypalon
    • Resistant to abrasion, ozone and oxidizing chemicals
    • 15 mil thickness
  • Butyl - Available in sizes 8.5 and 9.75
    • Provides high impermeability to water vapor, gases and toxic chemicals
    • Resistant to oxygenated solvents and most oxidizing chemicals
    • 15 mil thickness

Glove Box Sleeves

  • Neoprene Straight Sleeves
    • For use with Neoprene or Latex Hands
    • With O-rings and springs

Glove Box Hands - For use with neoprene straight or accordian sleeves

  • Neoprene available in size 7.5, 9 or 10
    • 17 mil thickness
    • Lightweight and embossed with firm hold finish
  • Latex available in size 7 or 9
    • 9 mil thickness
    • White, surgical-type

Specifications

Color Black
Hand Ambidextrous
Latex Free Yes
Material Neoprene
Product Type Glove Box Gloves
Size 7.5
Sterility Non-sterile
Thickness (English) 17 mil
Manufacturing/QMS Audit Standards ISO 9001
Model 5006000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.