Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gloveworks Black Nitrile Industrial Grade Gloves
GREENER_CHOICE

Catalog No. p-7219114
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Black nitrile industrial gloves offer the puncture resistance of latex, and provide protection against common and specialty chemicals. Micro-roughened surface and textured fingertips enhance grip, while the polymer coated interior leads to easier glove changes.

  • Constructed from medium-weight premium nitrile
  • At 5 mils thick, they are more than 50% thicker than standard nitrile gloves
  • Nitrile has three times the puncture resistance of latex
  • Strong barrier protection against common and specialty chemicals, including cleaning solutions
  • Micro-roughened surface and textured fingertips enhance grip
  • Polymer coated on the inside for easier glove changes
  • Black industrial gloves look professional while concealing dirt and grime
  • Ideal for professionals in automotive, industrial, janitorial, sanitation, food service, and safety applications
  • Latex-free for those allergic to natural rubber

Specifications

Applications Industrial
Color Black
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 6 mil
Finger Thickness (Metric) 0.15 mm
Laboratory/Cleanroom Use No
Latex Free Yes
Material Nitrile
Palm Thickness (English) 5 mil
Palm Thickness (Metric) 0.13 mm
Size Small
Series GPNB
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.