Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Harris Industries Glow Brite Glow-in-the-Dark Safety-Grade Tape
$34.04
Specifications
Description | Safety |
---|---|
Color | Ivory |
Format | Tape |
For Use With (Application) | Switches handrails building exits, hazardous areas uneven surfaces, low overheads, emergency equipment, stairways/doorways, fire extinguishers |
Catalog Number | Mfr. No. | Product Type | Size (English) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Size (English) | Quantity | Price | Quantity & Availability | |||
19-019992
![]() |
Harris Industries
GL002 |
Glow-in-the-Dark Tape | 2 in. x 10 yd. | 1 Ea. |
Each of 1 for $34.04
|
|
|||
Specifications
Safety | |
Ivory | |
Tape | |
Switches handrails building exits, hazardous areas uneven surfaces, low overheads, emergency equipment, stairways/doorways, fire extinguishers |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title