Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8915BK Class E Black Bottom Rain Pants

Catalog No. p-7163125
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
4X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Class E Black Bottom Rain Pants offer breathable, waterproof, comfort with black accents to hide dirt.

  • Weatherproof 300D oxford outer shell
  • Breathable PU coating
  • 2 in. ANSI-compliant 3M™ Scotchlite™ Reflective
  • Elastic waistband with drawstring
  • Zippered leg openings for easy on/off
  • One right leg cargo pocket
  • Black accents on bottom of leg

Specifications

Size Small
Color Yellow
Waist Style Elastic with drawstring
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.