Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLP-1R Antibody (7H7), Novus Biologicals™

Human Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | GLP-1R |
---|---|
Clone | 7H7 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
GLP-1R Monoclonal antibody specifically detects GLP-1R in Human samples. It is validated for ELISASpecifications
GLP-1R | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
GLP-1 receptor, GLP-1R, GLP-1-R, glucagon-like peptide 1 receptor, MGC138331 | |
Recombinant Human GLP-1R protein [UniProt P43220] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
7H7 | |
ELISA | |
Unconjugated | |
Human | |
Cancer, GPCR, Lipid and Metabolism, Signal Transduction | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
2740 | |
IgG4 | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title