Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLP-1R Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP197308AF594
Description
GLP-1R Polyclonal specifically detects GLP-1R in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
GLP-1R | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated | |
GLP-1 receptor, GLP-1R, GLP-1-R, glucagon-like peptide 1 receptor, MGC138331 | |
Rabbit | |
0.1 mL | |
2740 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
GLP1R | |
A synthetic peptide made to an internal portion of the human GLP1R protein (between residues 250-350) [UniProt P43220] | |
Primary | |
Human, Mouse, Rat, Canine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction