Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLP-1R Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NLS1205G
Description
GLP-1R Polyclonal specifically detects GLP-1R in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| GLP-1R | |
| Polyclonal | |
| Western Blot 1.0 μg/mL, Immunohistochemistry 10-20 μg/mL, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 10-20 μg/mL | |
| P43220 | |
| GLP1R | |
| A synthetic peptide made to an N-terminal extracellular portion of the human GLP1R protein (between residues 100-200). [UniProt P43220] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and mouse. The immunogen used for this antibody has 100/88% homology/identity with the rat protein. | |
| Store at 4C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GLP-1 receptor, GLP-1R, GLP-1-R, glucagon-like peptide 1 receptor, MGC138331 | |
| Rabbit | |
| 53 kDa | |
| 0.05 mL | |
| Cancer, GPCR, Lipid and Metabolism, Signal Transduction | |
| 2740 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction