Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLP-2R Antibody (413801), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4285B
Description
GLP-2R Monoclonal antibody specifically detects GLP-2R in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
GLP-2R | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
413801 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
GLP-2 receptor, GLP-2R, GLP-2-R, glucagon-like peptide 2 receptor | |
HEK293 human embryonic kidney cell line transfected with human GLP-2R, Met1-Ile553, Accession # O95838 | |
0.1 mL | |
Cell Cycle and Replication, GPCR | |
9340 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction