Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLP/EHMT1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP177400SS
Description
GLP/EHMT1 Polyclonal specifically detects GLP/EHMT1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
GLP/EHMT1 | |
Polyclonal | |
Unconjugated | |
PBS and 30% Glycerol with 0.05% Sodium Azide | |
EC 2.1.1.-, EC 2.1.1.43, euchromatic histone-lysine N-methyltransferase 1H3-K9-HMTase 5, Eu-HMTase1DEL9q34, EUHMTASE1FP13812, FLJ12879, G9a-like protein 1, GLP, GLP1, H3 lysine-9 specific 5, KIAA1876euchromatic histone methyltransferase 1, KMT1DDKFZp667M072 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
0.4 mg/mL | |
Western Blot reported in scientific literature (PMID 30858340), Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin 1:150, KnockDown Validated reported in scientific literature (PMID 30858340) | |
Q9H9B1 | |
EHMT1 | |
A genomic peptide made to an N-terminal region of the human EHMT1 protein (within residues 20-200). [Swiss-Prot Q9H9B1] | |
0.025 mL | |
Breast Cancer | |
79813 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction