Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLP Mouse anti-Human, Clone: 1G9, Bioss
SDP

Catalog No. 501979873
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9873 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9873 Supplier Bioss Supplier No. BSM0933M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EHMT1, also known as EUHMTASE1, is a histone methyltransferase. This protein methylates ′Lys-9′ of histone H3 in vitro. H3 ′Lys-9′ methylation represents a specific tag for epigenetic transcriptional repression by recruiting HP1 proteins to methylated histones. EHMT1 is Probably targeted to histone H3 by different DNA-binding proteins like E2F6, MGA, MAX and/or DP1. During G0 phase, it probably contributes to silencing of MYC- and E2F-responsive genes, suggesting a role in the G0/G1 transition of the cell cycle.

Specifications

Antigen GLP
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1G9
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene EHMT1
Gene Accession No. Q9H9B1
Gene Alias bA188C12.1; EHMT1; euchromatic histone lysine methyltransferase 1; Euchromatic histone-lysine N-methyltransferase 1; EUHMTASE1; Eu-HMTase1; FP13812; G9a-like protein 1; GLP; GLP1; H3-K9-HMTase 5; Histone H3-K9 methyltransferase 5; Histone-lysine N-methyltransferase EHMT1; histone-lysine N-methyltransferase, H3 lysine-9 specific 5; KIAA1876; KMT1D; Lysine N-methyltransferase 1D
Gene Symbols EHMT1
Host Species Mouse
Immunogen KLH conjugated synthetic peptide derived from human GLP-1.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79813
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.