Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLP1 Antibody (10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $560.50
Specifications
Antigen | GLP1 |
---|---|
Clone | 10 |
Concentration | 1.0 mg/mL |
Dilution | ELISA 1:20000 |
Applications | ELISA |
Description
GLP1 Monoclonal specifically detects GLP1 in Mammal samples. It is validated for Western Blot, ELISA.Specifications
GLP1 | |
1.0 mg/mL | |
ELISA | |
Unconjugated | |
Mouse | |
Cancer, GPCR, Signal Transduction | |
P01275 | |
2641 | |
Synthetic GLP-1(7-17) adsorbed onto aluminum hydroxide gel | |
Primary | |
Store at 4C in the dark. |
10 | |
ELISA 1:20000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
glicentin-related polypeptide, GLP1, glucagon-like peptide 1, GRPP | |
GCG | |
IgG2a κ | |
Protein A or G purified | |
This antibody binds the free N-terminus of GLP-1(7-37) and GLP-1(7-36)amide and shows <0.2% cross-reactivity with GLP-1(9-36)amide and human GIP. It ross-reacts 0.25% with GLP-1(1-37), approximately 6% with human GLP-2 and approximately 50% with exendin-4. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title