Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLP1 Antibody (10), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105180R
Description
GLP1 Monoclonal specifically detects GLP1 in Mammal samples. It is validated for Western Blot, ELISA.Specifications
GLP1 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GCG | |
Synthetic GLP-1(7-17) adsorbed onto aluminum hydroxide gel | |
0.1 mL | |
Cancer, GPCR, Signal Transduction | |
2641 | |
Human | |
Purified |
Western Blot, ELISA | |
10 | |
Western Blot, ELISA | |
glicentin-related polypeptide, GLP1, glucagon-like peptide 1, GRPP | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
This antibody binds the free N-terminus of GLP-1(7-37) and GLP-1(7-36)amide and shows <0.2% cross-reactivity with GLP-1(9-36)amide and human GIP. It ross-reacts 0.25% with GLP-1(1-37), approximately 6% with human GLP-2 and approximately 50% with exendin-4. | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction