Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLP1 Antibody (1D10), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32655850UL
Description
GLP1 Monoclonal antibody specifically detects GLP1 in Human samples. It is validated for ELISASpecifications
GLP1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
1D10 | |
ELISA | |
glicentin-related polypeptide, GLP1, glucagon-like peptide 1, GRPP | |
A synthesized peptide derived from Human GLP1 [UniProt P01275] | |
50 μL | |
Cancer, Signal Transduction, Stem Cell Markers | |
2641 | |
Store at -20 to -70C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction