Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLP1 Antibody (4), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP223558AF750
Description
GLP1 Monoclonal specifically detects GLP1 in Human, Mouse, Mammal samples. It is validated for ELISA, Immunohistochemistry.Specifications
| GLP1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| GCG | |
| Synthetic GLP-1(7-17) adsorbed onto aluminum hydroxide gel | |
| Primary | |
| This antibody binds the free N-terminus of GLP-1(7-37) and GLP-1(7-36)amide and shows <0.2% cross-reactivity with GLP-1(1-37), GLP-1(9-36)amide, human GIP and exendin-4. Cross-reacts approximately 1% with human GLP-2. | |
| Store at 4°C in the dark. |
| ELISA, Immunohistochemistry | |
| 4 | |
| ELISA, Immunohistochemistry | |
| glicentin-related polypeptide, GLP1, glucagon-like peptide 1, GRPP | |
| Mouse | |
| 0.1 mL | |
| 2641 | |
| Human, Mouse, Mammalia | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction