Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GLP1R Recombinant Human Monoclonal Antibody (7H7)
Human Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA550087
Description
GLP1R Recombinant Monoclonal Antibody for Flow, ELISA
Glucagon-like peptide-1(9-36)amide and GLP-1(9-37) are the forms of GLP-1 that result from the rapid degradation of the active forms of the peptide (GLP-1(7-36)amide and GLP-1(7-37)) by the enzyme dipeptidyl peptidase-IV (DPP-IV, also known as CD26 or adenosine deaminase binding protein). GLP-1 is a peptide hormone of the glucagon family, produced by the L cells of the intestinal mucosa from the same prohormone as glucagon. The active forms are potent stimulators of glucose-dependent insulin secretion. The sequence of GLP-1 is fully conserved in all mammalian species examined so far.
Specifications
GLP1R | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P43220 | |
Glp1r | |
Recombinant Human GLP1R protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG4 |
ELISA, Flow Cytometry, Western Blot | |
7H7 | |
Unconjugated | |
Glp1r | |
Glip; GLP-1; GLP1 receptor; GLP-1 receptor; GLP1R; GLP-1R; GLP-1-R; GLP1Rc; GLP-I receptor; Glpr; glucagon like peptide 1 receptor; glucagon-like peptide 1 receptor; glucagon-like peptide-1 receptor; LOW QUALITY PROTEIN: glucagon-like peptide 1 receptor; glucagon-like peptide 1 receptor; pancreatic beta cell receptor for the gluco-incretin hormone glucagon-like peptide 1; RATGL1RCP; seven transmembrane helix receptor | |
Human | |
Affinity Chromatography | |
RUO | |
2740 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction