Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GLP7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119695
This item is not returnable.
View return policy
Description
Positive Control: A. thaliana tissue. Subcellular Location: Apoplast.
May play a role in plant defense. Probably has no oxalate oxidase activity even if the active site is conserved.
Specifications
| GLP7 | |
| Polyclonal | |
| Unconjugated | |
| GLP7 | |
| Germin-like protein subfamily 1 member 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 837586 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| 0 | |
| GLP7 | |
| Recombinant protein within A. thaliana AtGLP7 aa 10-217. | |
| 100 μL | |
| Primary | |
| Plant | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction