Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLRA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521673
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521673 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521673 Supplier Invitrogen™ Supplier No. PA521673
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HepG2, mouse brain. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (95%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Glycine is an inhibitory neurotransmitter in the central nervous system. The glycine receptor (GlyR) is a member of the nicotinic acetylcholine receptor of ligand-gated ion channels. This receptor has important roles in a variety of physiological processes, especially in mediating inhibitory neurotransmission in the spinal cord and brain stem. GlyR is a pentameric receptor comprised of alpha and beta subunits. Four alpha-subunits (GLRA1, GLRA2, GLRA3, GLRA4) and one beta-subunit (GLRB) of GlyR have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLRA2
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene GLRA2
Gene Accession No. P23416, Q7TNC8
Gene Alias GLR; GLRA2; glycine receptor; glycine receptor alpha 2; glycine receptor alpha 2 subunit; Glycine receptor alpha 2 subunit (glycine receptor, neonatal); glycine receptor strychnine-binding subunit; Glycine receptor subunit alpha-2; glycine receptor, alpha 2; glycine receptor, alpha 2 subunit; glycine receptor, alpha-2 polypeptide; RNNEOGLY
Gene Symbols GLRA2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Extracellular domain of human Glycine Receptor alpha 2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 237213, 2742
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.