Learn More
Invitrogen™ GLRA4 Polyclonal Antibody

Description
Immunogen sequence: IRLRRRQRRQ RLEEDIIQES RFYFRGYGLG HCLQARDGGP MEGSGIYSPQ PPAPLLREGE TTRKLYVD Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 81%.
Specifications
Specifications
| Antigen | GLRA4 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | GLRA4 |
| Gene Accession No. | 0 |
| Gene Alias | Glra4; glycine receptor alpha 4; glycine receptor subunit alpha-4; glycine receptor, alpha 4; glycine receptor, alpha 4 subunit; RGD1564103 |
| Gene Symbols | GLRA4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.