Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLRB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101610
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101610 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101610 Supplier Invitrogen™ Supplier No. PA5101610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GLRB.

In the central nervous system (CNS), glycine-mediated inhibitory neurotransmission is essential to voluntary motor control and reflex responses. Glycine binds to glycine receptors (GlyR) in the postsynaptic neuronal membranes. GlyR, gamma-aminobutryic acid, serotonin and acetylcholine comprise an evolutionally conserved superfamily of ligand-gated ion channels. The pentameric subunit structure of GlyR consists of two types of glycosylated membrane proteins, alpha1 through alpha4 and beta, and an associated peripheral membrane protein, which combine to form a chloride-selective ion channel. In humans, the composition of the pentamer changes from alpha2 subunits in the fetal CNS to alpha1 and beta subunits in the adult CNS. Fast potentiation of GlyR by intracellular Ca2+ in the brainstem and midbrain indicate an important role for Ca2+ in modulation of glycinergic synapses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLRB
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GLRB
Gene Accession No. P20781, P48167, P48168
Gene Alias AI853901; Glrb; glycine receptor; Glycine receptor 58 kDa subunit; glycine receptor beta; glycine receptor subunit beta; glycine receptor, beta; glycine receptor, beta subunit; GlyR beta; Glyrb; HKPX2; inhibitory glycine receptor; spa; spastic
Gene Symbols GLRB
Host Species Rabbit
Immunogen A synthesized peptide derived from human GLRB(Accession P48167), corresponding to amino acid residues D210-F260.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14658, 25456, 2743
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.