Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GLRB Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112268
Description
GLRB Polyclonal Antibody for Western Blot, IHC (P), IP
In the central nervous system (CNS), glycine-mediated inhibitory neurotransmission is essential to voluntary motor control and reflex responses. Glycine binds to glycine receptors (GlyR) in the postsynaptic neuronal membranes. GlyR, gamma-aminobutryic acid, serotonin and acetylcholine comprise an evolutionally conserved superfamily of ligand-gated ion channels. The pentameric subunit structure of GlyR consists of two types of glycosylated membrane proteins, alpha1 through alpha4 and beta, and an associated peripheral membrane protein, which combine to form a chloride-selective ion channel. In humans, the composition of the pentamer changes from alpha2 subunits in the fetal CNS to alpha1 and beta subunits in the adult CNS. Fast potentiation of GlyR by intracellular Ca2+ in the brainstem and midbrain indicate an important role for Ca2+ in modulation of glycinergic synapses.
Specifications
| GLRB | |
| Polyclonal | |
| Unconjugated | |
| GLRB | |
| AI853901; Glrb; glycine receptor; Glycine receptor 58 kDa subunit; glycine receptor beta; glycine receptor subunit beta; glycine receptor, beta; glycine receptor, beta subunit; GlyR beta; Glyrb; HKPX2; inhibitory glycine receptor; spa; spastic | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 2743 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| P48167 | |
| GLRB | |
| E. coli-derived Human GLRB fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction