Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLRX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544837
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544837 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544837 Supplier Invitrogen™ Supplier No. PA544837
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: NVNYKVVELD LLEYGNQFQD ALYKMTGERT VPRIFVNGTF IGGATDTHRL H Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 86%; Sheep: 86%.

Glutaredoxin (Grx), also known as thiol transferase, is a small heat-stable oxidoreductase. Grxs form part of the glutaredoxin system, comprising NADPH, GSH and glutathione reductase, which transfers electrons from NADPH to glutaredoxins via GSH. First discovered in E. coli as GSH-dependent hydrogen donors for ribonucleotide reductase, Grx catalyzes GSH-disulfide oxidoreductase via two redox-active cysteine residues. The active sequence (Cys-Pro-Tyr-Cys) is conserved in a variety of species. The 12-kD dithiol protein has a role in reduction of mixed disulfides in cells exposed to oxidative stress.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLRX2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GLRX2
Gene Accession No. Q9NS18
Gene Alias 1700010P22Rik; AI645710; bA101E13.1 (GRX2 glutaredoxin (thioltransferase) 2); CGI-133; GLRX2; glutaredoxin (thioltransferase) 2; glutaredoxin 2; glutaredoxin 2 (thioltransferase); glutaredoxin-2, mitochondrial; GRX2
Gene Symbols GLRX2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human GLRX2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51022
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.