Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLRX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554692
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554692 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554692 Supplier Invitrogen™ Supplier No. PA554692
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DMNVNYKVVE LDLLEYGNQF QDALYKMTGE RTVPRIFVNG TFIGGATDTH RLHKEGKLLP LVHQCYL Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.

Glutaredoxin (Grx), also known as thiol transferase, is a small heat-stable oxidoreductase. Grxs form part of the glutaredoxin system, comprising NADPH, GSH and glutathione reductase, which transfers electrons from NADPH to glutaredoxins via GSH. First discovered in E. coli as GSH-dependent hydrogen donors for ribonucleotide reductase, Grx catalyzes GSH-disulfide oxidoreductase via two redox-active cysteine residues. The active sequence (Cys-Pro-Tyr-Cys) is conserved in a variety of species. The 12-kD dithiol protein has a role in reduction of mixed disulfides in cells exposed to oxidative stress.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLRX2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GLRX2
Gene Accession No. Q9NS18
Gene Alias 1700010P22Rik; AI645710; bA101E13.1 (GRX2 glutaredoxin (thioltransferase) 2); CGI-133; GLRX2; glutaredoxin (thioltransferase) 2; glutaredoxin 2; glutaredoxin 2 (thioltransferase); glutaredoxin-2, mitochondrial; GRX2
Gene Symbols GLRX2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GLRX2. Recombinant protein control fragment (Product #RP-92075).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51022
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.