Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLRX5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559985
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559985 Supplier Invitrogen™ Supplier No. PA559985
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVKKDKVVVF LKGTPEQPQC GFSNAVVQIL RLHGVRDYAA YNVLDDPELR QGIKDYSNWP TIPQVYLNGE FVGGCDILLQ MHQNGDLV Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.

GLRX5 is monothiol glutaredoxin involved in the biogenesis of iron-sulfur clusters. GLRX5 required for normal iron homeostasis. GLRX5 is required for normal regulation of hemoglobin synthesis by the iron-sulfur protein ACO1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLRX5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GLRX5
Gene Accession No. Q86SX6
Gene Alias 2310004O13Rik; 2900070E19Rik; AU020725; C14orf87; FLB4739; GLRX5; glutaredoxin 5; glutaredoxin 5 homolog; glutaredoxin 5 homolog (S. cerevisiae); glutaredoxin-related protein 5, mitochondrial; GRX5; Monothiol glutaredoxin-5; PR01238; PRO1238; PRSA; RGD1308383; SIDBA3; SPAHGC
Gene Symbols GLRX5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GLRX5. Recombinant protein control fragment (Product #RP-99951).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51218
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.